| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000547951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000547951.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552237.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000547951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000547951.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000549639.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000551837.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552237.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
ENST00000552440.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011588.hg.1 |
NR_125360.2 |
Refseq |