Search
Search

SMCR2| Gene Symbol: SMCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131243.1 | 3 | 49 | |||
| GenBank mRNA | AI821738.1 | - | 2 | 253 | 49 | |
| AI821758.1 | - | 2 | 252 | 49 | ||
| BX116483.1 | - | 2 | 263 | 49 | ||
Set Membership: |
|