Search
Search

C19orf68| Gene Symbol: C19orf68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199341.3 | NP_955373.3 | 4 | 2046 | 108 | |
| XM_005259449.3 | XP_005259506.1 | 4 | 2484 | 108 | ||
| XM_006723204.3 | XP_006723267.1 | 4 | 2181 | 108 | ||
| XM_006723205.2 | XP_006723268.1 | 4 | 2072 | 108 | ||
| XM_011526936.2 | XP_011525238.1 | 3 | 2086 | 108 | ||
| XM_011526937.2 | XP_011525239.1 | 2 | 1588 | 108 | ||
Set Membership: |
|