Search
Search

RASD2| Gene Symbol: RASD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014310.3 | NP_055125.2 | 3 | 1705 | 103 | |
| XM_005261442.4 | XP_005261499.1 | 3 | 1565 | 103 | ||
| XM_011530040.2 | XP_011528342.1 | 4 | 2536 | 103 | ||
| XM_017028701.1 | XP_016884190.1 | 2 | 4314 | 103 | ||
| XM_017028702.1 | XP_016884191.1 | 3 | 2623 | 103 | ||
| GenBank mRNA | AF279143.1 | - | 2 | 1536 | 103 | |
| AL831936.1 | - | 2 | 1414 | 103 | ||
| BC013419.2 | - | 3 | 1707 | 3604196 | 103 | |
Set Membership: |
|