Search
Search

MANEAL| Gene Symbol: MANEAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031740.2 | NP_001026910.1 | 4 | 1613 | 110 | |
| NM_001113482.1 | NP_001106954.1 | 4 | 1890 | 110 | ||
| NM_152496.2 | NP_689709.1 | 2 | 1431 | 110 | ||
| XM_005270510.3 | XP_005270567.1 | 3 | 1822 | 110 | ||
| XM_011540788.1 | XP_011539090.1 | 3 | 1432 | 110 | ||
| GenBank mRNA | AB188490.1 | - | 4 | 1832 | 110 | |
| AK055996.1 | - | 2 | 1431 | 110 | ||
| BC009952.2 | - | 2 | 835 | 110 | ||
| BC031903.1 | - | 1 | 821 | 110 | ||
| BC038190.1 | - | 3 | 566 | 110 | ||
| BC063587.1 | - | 4 | 1347 | 110 | ||
| BC077730.1 | - | 4 | 1613 | 6265730 | 110 | |
Set Membership: |
|