Search
Search

ZNF517| Gene Symbol: ZNF517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317936.1 | NP_001304865.1 | 4 - 5 | 364 | 88 | |
| NM_213605.2 | NP_998770.2 | 4 - 5 | 385 | 88 | ||
| NR_134284.1 | 4 - 5 | 385 | 88 | |||
| XM_011517014.2 | XP_011515316.1 | 4 - 5 | 924 | 88 | ||
| XM_011517015.2 | XP_011515317.1 | 4 - 5 | 366 | 88 | ||
| XM_011517017.2 | XP_011515319.1 | 3 - 4 | 783 | 88 | ||
| GenBank mRNA | AK096527.1 | - | 4 - 5 | 924 | 88 | |
| AK097278.1 | - | 2 - 3 | 1408 | 88 | ||
| AK131440.1 | - | 4 - 5 | 385 | 88 | ||
| BC126161.1 | - | 3 - 4 | 305 | 8991877 | 88 | |
| BC143922.1 | - | 2 - 3 | 178 | 88 | ||
| DB016003.1 | - | 4 - 5 | 385 | 88 | ||
| DB165783.1 | - | 4 - 5 | 323 | 88 | ||
Set Membership: |
|