Search
Search

SIRT6| Gene Symbol: SIRT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193285.2 | NP_001180214.1 | 4 - 5 | 501 | 69 | |
| NM_001321058.1 | NP_001307987.1 | 3 - 4 | 373 | 69 | ||
| NM_001321059.1 | NP_001307988.1 | 3 - 4 | 318 | 69 | ||
| NM_001321060.1 | NP_001307989.1 | 4 - 5 | 501 | 69 | ||
| NM_001321061.1 | NP_001307990.1 | 3 - 4 | 373 | 69 | ||
| NM_001321062.1 | NP_001307991.1 | 2 - 3 | 190 | 69 | ||
| NM_001321063.1 | NP_001307992.1 | 3 - 4 | 318 | 69 | ||
| NM_001321064.1 | NP_001307993.1 | 4 - 5 | 466 | 69 | ||
| NM_016539.3 | NP_057623.2 | 4 - 5 | 501 | 69 | ||
| GenBank mRNA | AF233396.1 | - | 4 - 5 | 496 | 69 | |
| AK074810.1 | - | 4 - 5 | 464 | 69 | ||
| AK225567.1 | - | 4 - 5 | 469 | 69 | ||
| AK293348.1 | - | 3 - 4 | 334 | 69 | ||
| AK299420.1 | - | 3 - 4 | 316 | 69 | ||
| AK301247.1 | - | 3 - 4 | 281 | 69 | ||
| AK315048.1 | - | 4 - 5 | 465 | 69 | ||
| AL559383.3 | - | 4 - 5 | 439 | 69 | ||
| BC004218.2 | - | 3 - 4 | 313 | 3352203 | 69 | |
| BC005026.1 | - | 4 - 5 | 439 | 2820472 | 69 | |
| BC028220.1 | - | 4 - 5 | 447 | 5217599 | 69 | |
| BE281103.1 | - | 3 - 4 | 299 | 69 | ||
| BP418952.1 | - | 2 - 3 | 135 | 69 | ||
| BQ060703.1 | - | 3 - 4 | 214 | 69 | ||
| CN364175.1 | - | 4 - 5 | 444 | 69 | ||
| CN364177.1 | - | 4 - 5 | 504 | 69 | ||
| CR457200.1 | - | 4 - 5 | 436 | 69 | ||
Set Membership: |
|