Search
Search
| Gene Symbol: FMN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305424.2 | NP_001292353.1 | 8 - 9 | 4399 | 92 | |
| NM_001348094.2 | NP_001335023.1 | 5 - 6 | 2308 | 92 | ||
| NM_020066.5 | NP_064450.3 | 7 - 8 | 4387 | 92 | ||
| XM_017001837.2 | XP_016857326.1 | 8 - 9 | 4399 | 92 | ||
| XM_017001840.3 | XP_016857329.1 | 7 - 8 | 2637 | 92 | ||
| XM_017001841.3 | XP_016857330.1 | 7 - 8 | 3095 | 92 | ||
| GenBank mRNA | AB209153.1 | - | 8 - 9 | 2990 | 92 | |
| AK297716.1 | - | 2 - 3 | 155 | 92 | ||
| AK297755.1 | - | 2 - 3 | 401 | 92 | ||
| DN990332.1 | - | 3 - 4 | 389 | 92 | ||
Set Membership: |
|