Search
Search

CCDC179| Gene Symbol: CCDC179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195637.1 | NP_001182566.1 | 1 - 2 | 46 | 73 | |
| XM_011519802.2 | XP_011518104.1 | 1 - 2 | 591 | 73 | ||
| XM_011519803.2 | XP_011518105.1 | 1 - 2 | 579 | 73 | ||
| GenBank mRNA | AI651461.1 | - | 3 - 4 | 320 | 73 | |
Set Membership: |
|