Search
Search
| Gene Symbol: LLPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032338.3 | NP_115714.1 | 2 - 3 | 272 | 126 | |
| GenBank mRNA | AK057947.1 | - | 1 - 2 | 185 | 126 | |
| AK091568.1 | - | 2 - 3 | 272 | 126 | ||
| BC006002.1 | - | 2 - 3 | 268 | 4247279 | 126 | |
| BC070232.1 | - | 2 - 3 | 264 | 4594274 | 126 | |
| BC107780.1 | - | 2 - 3 | 6698895 | 126 | ||
Set Membership: |
|