Search
Search

ZNF765| Gene Symbol: ZNF765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040185.3 | NP_001035275.1 | 4 | 2621 | 126 | |
| NM_001350495.2 | NP_001337424.1 | 3 | 2494 | 126 | ||
| GenBank mRNA | AK293890.1 | - | 6 | 2842 | 126 | |
| BC140724.1 | - | 4 | 2602 | 126 | ||
| BX647981.1 | - | 4 | 2620 | 126 | ||
Set Membership: |
|