Search
Search

HMGCS2| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542142.2 | XP_005542199.1 | 3 - 4 | 953 | 129 | |
| XM_005542143.2 | XP_005542200.1 | 3 - 4 | 732 | 129 | ||
Set Membership: |
|