Search
Search

Vegfc| Gene Symbol: Vegfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009506.2 | NP_033532.1 | 1 - 2 | 391 | 78 | |
| GenBank mRNA | AK047844.1 | - | 318 | 78 | ||
| BC096377.1 | - | 1 - 2 | 296 | 5002720 | 78 | |
| EU862554.1 | - | 1 - 2 | 144 | 78 | ||
| EU862555.1 | - | 1 - 2 | 144 | 78 | ||
| U58112.1 | - | 1 - 2 | 287 | 78 | ||
| U73620.1 | - | 1 - 2 | 297 | 78 | ||
Set Membership: |
|