Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010588.2 | NP_034718.2 | 4 - 5 | 742 | 61 | |
GenBank mRNA | AF038572.1 | - | 4 - 5 | 742 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |