| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10569396 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10569395 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10569394 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10569397 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10569398 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1418802_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10569393 |
- |
- |
| Mouse Genome 430 2.0 Array |
1418802_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1418802_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
97423_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
97423_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700038635.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038635.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038635.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038636.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038636.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038636.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038641.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038641.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038643.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038643.mm.1 |
NR_110354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038643.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038643.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038634.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038634.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038634.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038638.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038638.mm.1 |
NR_110354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038638.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038638.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038639.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038639.mm.1 |
NR_110354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038639.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038639.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038642.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038642.mm.1 |
NR_110354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038642.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038642.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038645.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038645.mm.1 |
NR_110354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038645.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038645.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038644.mm.1 |
NR_015529.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038644.mm.1 |
NR_110354.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038644.mm.1 |
NR_110355.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038644.mm.1 |
TC0700004578.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1418802_PM_at |
- |
- |
| Mouse Expression Set 430 |
1418802_at |
- |
- |
| Murine Genome U74v2 Set |
161058_PM_f_at |
- |
- |
| Murine Genome U74v2 Set |
161058_f_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1418802_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1418802_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1418802_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1418802_PM_at |
- |
- |