| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26229206 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26229204 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26229207 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26229202 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1986158 |
BC058359.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1986158 |
BC012312.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1986158 |
BC023738.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1986158 |
AK089954.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1986158 |
AK161674.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
224072 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
909133 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
237937 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
797921 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
75539 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693264 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729180 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
263871 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
AK088020.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
AK088079.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
AK132250.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
AK137794.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
BC011513.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
M12848.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
M13138.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
M16449.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
NM_010848.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
455721 |
X02774.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
508096 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839276 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
844868 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
708775 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
424171 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
269590 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636332 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
293664 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
69675 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
17172 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
16021 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
95906 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
139702 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P163806 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P106070 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1986158 |
BC058359.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1986158 |
BC012312.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1986158 |
BC023738.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1986158 |
AK089954.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1986158 |
AK161674.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229213 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229204 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229227 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229211 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229202 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229216 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229215 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229203 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229200 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229207 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229198 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229208 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229218 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229222 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229223 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229209 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229197 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229225 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26229205 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06667828 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P163806 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06667797 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01847415 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P106070 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P153967 |
BC058359.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P153967 |
BC012312.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P153967 |
BC023738.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P153967 |
AK089954.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P153967 |
AK161674.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130965 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P163806 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P106070 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07303022 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07303016 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07303018 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07303028 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07303021 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07303017 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07303029 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26229202 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26229207 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26229206 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26229204 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229227 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228972 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229098 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229030 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228978 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229021 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229115 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229043 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229011 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229145 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229023 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228997 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229046 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229120 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229089 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228987 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229216 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229071 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229058 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229099 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228969 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229074 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229052 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229056 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229080 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229050 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229093 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229218 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229051 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228974 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229087 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229221 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229131 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229124 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229204 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229141 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228981 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229148 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228971 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228975 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229108 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229209 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229042 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229036 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229133 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229101 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228976 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229082 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229197 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229130 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229054 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229057 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229213 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229083 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229075 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229003 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229138 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229091 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228967 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229008 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229014 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229225 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229073 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229104 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229103 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26228984 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229059 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26229045 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229223 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229221 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229203 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229213 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229218 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229204 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229202 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229227 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229208 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07276520 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06694816 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07303016 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229215 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07276495 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229205 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229211 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229225 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229216 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06876446 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229207 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26229197 |
- |
- |