| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
BC005481.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
AK140388.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
AK155486.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
AK088995.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
AK029337.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
BC005481.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
AK140388.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
AK155486.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
AK088995.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P222359 |
AK029337.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
214814 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
326817 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
550541 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583854 |
AK033294.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583854 |
AK047691.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583854 |
AK088218.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583854 |
AK141504.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583854 |
AK142228.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583854 |
AY266418.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583854 |
BC096540.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583854 |
NM_178909.4 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932656 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
11647 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P109338 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
BC005481.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
AK140388.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
AK155486.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
AK088995.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
AK029337.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
BC005481.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
AK140388.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
AK155486.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
AK088995.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P222359 |
AK029337.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
BC005481.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
AK140388.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
AK155486.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
AK088995.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
AK029337.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
BC005481.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
AK140388.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
AK155486.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
AK088995.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P222359 |
AK029337.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575436 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575466 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575427 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575441 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575464 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575471 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575456 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575450 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575449 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575460 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575426 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575459 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575432 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575455 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575425 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575457 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575444 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575445 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575447 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575423 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575446 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575421 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575419 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575451 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575462 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575452 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575465 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575472 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575431 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575428 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575433 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575429 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575417 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575468 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575438 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575440 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575467 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575442 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575435 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575454 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23575434 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05521465 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P109338 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P222359 |
BC005481.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
AK140388.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
AK155486.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
AK088995.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
AK029337.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
BC005481.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
AK140388.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
AK155486.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
AK088995.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P222359 |
AK029337.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P105377 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05521465 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P109338 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04263888 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04263879 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04698438 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04698444 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575464 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575417 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575437 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575535 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575445 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575473 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575364 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575414 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575385 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575472 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575375 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575545 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575401 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575479 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575518 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575447 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575380 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575520 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575428 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575368 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575431 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575465 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575446 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575519 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575438 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575495 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575373 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575462 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575534 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575526 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575426 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575402 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575423 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575409 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575501 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575366 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575386 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575444 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575531 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575485 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575451 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575392 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575411 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575507 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575508 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575497 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575467 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575459 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575381 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575416 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575372 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575388 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575387 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575514 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575523 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575533 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575505 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575475 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575425 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575410 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575516 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575454 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575538 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575513 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575528 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575511 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575395 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575397 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575419 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575432 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575384 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575504 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575460 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575541 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575405 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575456 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575499 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575457 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575449 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575450 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575483 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575435 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575509 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575421 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575492 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575389 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575442 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575529 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575427 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575471 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575506 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575468 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575407 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575522 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575542 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575477 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575489 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575466 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575452 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575429 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575548 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575413 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575503 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575378 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575482 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575390 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575543 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23575434 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575401 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575465 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575437 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575399 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04263887 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04177538 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575446 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575435 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575403 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575451 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575466 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575472 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575447 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575417 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04643977 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04698442 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575432 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575405 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575428 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575444 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575445 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04563190 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575413 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575402 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04643960 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575449 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575440 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575426 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575459 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575464 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04643951 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04263889 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575468 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04263891 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04643966 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04574344 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575442 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575431 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575471 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575419 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575395 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575429 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575434 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04973063 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575456 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575425 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575407 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575414 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04939922 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23575458 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04177550 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04698441 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04643955 |
- |
- |