Search
Search

Tfcp2l1| Gene Symbol: Tfcp2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023755.2 | NP_076244.2 | 1 - 2 | 159 | 78 | |
| GenBank mRNA | AF311309.1 | - | 1 - 2 | 154 | 78 | |
| AK007654.1 | - | 1 - 2 | 160 | 78 | ||
| AK143960.1 | - | 1 - 2 | 158 | 78 | ||
| BC116663.1 | - | 1 - 2 | 154 | 40087602 | 78 | |
| BY707692.1 | - | 1 - 2 | 160 | 78 | ||
Set Membership: |
|