Search
Search
| Gene Symbol: Nop9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026403.3 | NP_080679.3 | 5 - 6 | 1227 | 62 | |
| GenBank mRNA | AK011570.1 | - | 5 - 6 | 1217 | 62 | |
| AK015585.1 | - | 5 - 6 | 977 | 62 | ||
| AK032873.1 | - | 5 - 6 | 1211 | 62 | ||
| AK037831.1 | - | 5 - 6 | 1215 | 62 | ||
| AK050912.1 | - | 5 - 6 | 1218 | 62 | ||
| AK083861.1 | - | 5 - 6 | 1096 | 62 | ||
| AK139689.1 | - | 5 - 6 | 1178 | 62 | ||
| AK146020.1 | - | 5 - 6 | 1182 | 62 | ||
| AK146687.1 | - | 5 - 6 | 1199 | 62 | ||
| AK152732.1 | - | 5 - 6 | 1201 | 62 | ||
| AK154387.1 | - | 5 - 6 | 1219 | 62 | ||
| AK159798.1 | - | 5 - 6 | 1201 | 62 | ||
| AK178392.1 | - | 2 - 3 | 347 | 62 | ||
| BC018523.1 | - | 5 - 6 | 1207 | 3499986 | 62 | |
Set Membership: |
|