| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200010227.mm.1 |
NM_001252071.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010227.mm.1 |
NM_001252071.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010227.mm.1 |
NM_015764.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010227.mm.1 |
NM_015764.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010227.mm.1 |
TC1200001461.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010225.mm.1 |
NM_001252071.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010225.mm.1 |
NM_001252071.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010225.mm.1 |
NM_015764.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010225.mm.1 |
NM_015764.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010225.mm.1 |
TC1200001461.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010226.mm.1 |
NM_001252071.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010226.mm.1 |
NM_001252071.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010226.mm.1 |
NM_015764.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010226.mm.1 |
NM_015764.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010226.mm.1 |
TC1200001461.mm.1 |
MTA TC |