| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
AK019261.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
AK170168.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
BC027500.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
AK018003.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
NM_026077.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
AK019261.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
AK170168.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
BC027500.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
AK018003.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P263251 |
NM_026077.3 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
408941 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
23424 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
810576 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
409438 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453514 |
AK156682.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453514 |
BC026662.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453514 |
NM_152807.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
270662 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P139005 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
AK019261.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
AK170168.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
BC027500.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
AK018003.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
NM_026077.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
AK019261.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
AK170168.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
BC027500.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
AK018003.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P263251 |
NM_026077.3 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273995 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25274000 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273987 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273984 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273998 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273971 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273977 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273975 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25274001 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273992 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273982 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273988 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273974 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273986 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273976 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273970 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273991 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273997 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273990 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273981 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25273983 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P139005 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06259932 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P263251 |
AK019261.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
AK170168.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
BC027500.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
AK018003.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
NM_026077.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
AK019261.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
AK170168.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
BC027500.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
AK018003.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P263251 |
NM_026077.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121858 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P139005 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06064074 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06064077 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06064061 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06529406 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06064068 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273974 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273921 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25274000 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273907 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273923 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273964 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273977 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273950 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273987 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273946 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273918 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273966 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273993 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273934 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273992 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273954 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273967 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273930 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273917 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273932 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273955 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273981 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273975 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273953 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273997 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273959 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273984 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273903 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273983 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273986 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273944 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273922 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273971 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273909 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273905 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25273982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273967 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06351872 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273986 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273981 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273955 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06235769 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273968 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273917 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06063919 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273953 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06600873 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273991 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273992 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273954 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06600877 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273961 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273976 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273995 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273988 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273970 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273959 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273974 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273977 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273950 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273964 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06335095 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06412556 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273946 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273975 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273984 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273971 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273999 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273983 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273997 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25274001 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25273951 |
- |
- |