| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254631 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254629 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P401504 |
CJ312879.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P401504 |
BC029697.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P401504 |
NM_007741.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P401504 |
BB732275.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P401504 |
CJ312879.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P401504 |
BC029697.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P401504 |
NM_007741.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P401504 |
BB732275.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
503251 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616647 |
BC104128.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616647 |
BC104129.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616647 |
BC115846.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616647 |
BC127222.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616647 |
BC127223.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616647 |
NM_146306.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
153563 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
218122 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
681944 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
375584 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
845432 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914653 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
100559 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
313867 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
63033 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
121507 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P180050 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P152747 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P401504 |
CJ312879.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P401504 |
BC029697.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P401504 |
NM_007741.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P401504 |
BB732275.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P401504 |
CJ312879.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P401504 |
BC029697.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P401504 |
NM_007741.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P401504 |
BB732275.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401504 |
CJ312879.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401504 |
BC029697.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401504 |
NM_007741.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401504 |
BB732275.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401504 |
CJ312879.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401504 |
BC029697.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401504 |
NM_007741.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P401504 |
BB732275.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254621 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254618 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254612 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254641 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254615 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254620 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254643 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254640 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254634 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254646 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254610 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254629 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254619 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254638 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254609 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254635 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254636 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254632 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254626 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254613 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254648 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254624 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254607 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254627 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254631 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23254633 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P152747 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P401504 |
CJ312879.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P401504 |
BC029697.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P401504 |
NM_007741.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P401504 |
BB732275.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P401504 |
CJ312879.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P401504 |
BC029697.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P401504 |
NM_007741.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P401504 |
BB732275.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139614 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P152747 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03388958 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03413444 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03388960 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03413449 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03413443 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03413450 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03388961 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03413445 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254631 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23254629 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254713 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254700 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254679 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254715 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254606 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254698 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254748 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254738 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254719 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254662 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254594 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254654 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254665 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254737 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254656 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254597 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254706 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254664 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254613 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254693 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254600 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254646 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254657 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254618 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254712 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254682 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254672 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254674 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254675 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254649 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254631 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254744 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254684 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254627 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254609 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254729 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254632 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254653 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254735 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254708 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254648 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254670 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254731 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254651 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254661 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254697 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254680 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254701 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254722 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254641 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254725 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254634 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254668 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254658 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254734 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254629 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254619 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254717 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254720 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254724 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254636 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254635 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254667 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254709 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254705 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254607 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254721 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254677 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254595 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254714 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254716 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254660 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254643 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254727 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23254624 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03407102 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254656 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254627 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254682 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03413446 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254621 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254657 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254622 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254619 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254661 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254665 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254651 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03753013 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254687 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03413456 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254618 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03757458 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03753029 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03753025 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254658 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254660 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254683 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03676616 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03676621 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03590040 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03346677 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254601 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254624 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03413443 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254670 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03934912 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254610 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254648 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254634 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254654 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254629 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254633 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254664 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254646 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254674 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254643 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254640 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254679 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254638 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04081147 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03302999 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254688 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254607 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254684 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254641 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254675 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03339304 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254680 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03753021 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P23254667 |
- |
- |