Search
Search
| Gene Symbol: Gbas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008095.4 | NP_032121.3 | 7 - 8 | 675 | 67 | |
| GenBank mRNA | AJ001261.1 | - | 7 - 8 | 653 | 67 | |
| BC055893.1 | - | 7 - 8 | 624 | 4023066 | 67 | |
Set Membership: |
|