Search
Search
| Gene Symbol: Mcm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033196.1 | - | 9 - 10 | 1267 | 147 | |
| AK051795.1 | - | 9 - 10 | 1237 | 147 | ||
| AK088558.1 | - | 2 - 3 | 183 | 147 | ||
| AK133826.1 | - | 9 - 10 | 1268 | 147 | ||
| AK209639.1 | - | 1 - 2 | 123 | 147 | ||
| AK212108.1 | - | 1 - 2 | 123 | 147 | ||
| AK214383.1 | - | 1 - 2 | 123 | 147 | ||
| BC090645.1 | - | 9 - 10 | 1220 | 147 | ||
| BC094416.1 | - | 9 - 10 | 1232 | 5707724 | 147 | |
| D26090.1 | - | 8 - 9 | 1287 | 147 | ||
Set Membership: |
|