Search
Search

Arntl| Gene Symbol: Arntl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243048.1 | NP_001229977.1 | 5 - 6 | 612 | 69 | |
| NM_007489.4 | NP_031515.1 | 8 - 9 | 905 | 69 | ||
| GenBank mRNA | AB012601.1 | - | 5 - 6 | 406 | 69 | |
| AB012602.1 | - | 4 - 5 | 385 | 69 | ||
| AB014494.1 | - | 8 - 9 | 791 | 69 | ||
| AB015203.1 | - | 4 - 5 | 385 | 69 | ||
| AK143267.1 | - | 8 - 9 | 833 | 69 | ||
| AK146628.1 | - | 8 - 9 | 905 | 69 | ||
| AK147145.1 | - | 8 - 9 | 905 | 69 | ||
| AY690602.1 | - | 5 - 6 | 406 | 69 | ||
| BC011080.1 | - | 5 - 6 | 563 | 3661605 | 69 | |
| BC025973.1 | - | 6 - 7 | 656 | 3980719 | 69 | |
Set Membership: |
|