Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508100_m1
          See other TMEM176B GE Assays ›
          Gene Symbol:
          Tmem176b
          Gene Name:
          transmembrane protein 176B
          Gene Aliases:
          1810009M01Rik, Clast1, Lr8
          Chromosome Location:
          Chr.6: 48833811 - 48845950 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508100_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem176b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164207.1 NP_001157679.1 2 - 3 571 68
          NM_001164208.1 NP_001157680.1 2 - 3 219 68
          NM_001164209.2 NP_001157681.1 3 - 4 427 68
          NM_001286651.1 NP_001273580.1 3 - 4 424 68
          NM_001286652.1 NP_001273581.1 2 - 3 331 68
          NM_023056.4 NP_075543.1 2 - 3 820 68
          GenBank mRNA AB031386.1 - 2 - 3 380 68
          AK007408.1 - 2 - 3 226 68
          AK156042.1 - 2 - 3 572 68
          AK177944.1 - 1 - 2 125 68
          BC019539.1 - 1 - 2 189 4237639 68
          BC022166.1 - 2 - 3 835 3582123 68
          BQ571258.1 - 2 - 3 211 68
          CT010176.1 - 1 - 2 184 68
          CX213755.1 - 2 - 3 331 68
          DV047496.1 - 2 - 3 259 68
          DV059241.1 - 2 - 3 295 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002233.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem176b

          Gene Ontology Categories:

          Function(s) Process(es)

          cell differentiation
          negative regulation of dendritic cell differentiation
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 TC0600002233.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_001164207.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_001164207.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_001164208.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_001164208.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_001164209.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_001286651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_001286652.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_023056.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NM_023056.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017550.mm.1 NR_104575.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline