| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26022400 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26022398 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26022402 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
AK005954.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
AK082833.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
AK167332.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
NR_031759.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
BC094628.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
AK005954.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
AK082833.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
AK167332.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
NR_031759.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P273538 |
BC094628.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463624 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453736 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
533042 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
369283 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
324225 |
AK004033.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
324225 |
AK146097.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
324225 |
AK148270.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
324225 |
BC013766.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
324225 |
NM_008929.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
324225 |
U28423.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
275160 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
796146 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
918506 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
114715 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
5409 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P158677 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P146075 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
AK005954.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
AK082833.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
AK167332.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
NR_031759.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
BC094628.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
AK005954.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
AK082833.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
AK167332.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
NR_031759.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P273538 |
BC094628.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
AK005954.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
AK082833.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
AK167332.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
NR_031759.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
BC094628.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
AK005954.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
AK082833.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
AK167332.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
NR_031759.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P273538 |
BC094628.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022390 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022379 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022397 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022406 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022399 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022404 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022367 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022377 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022409 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022386 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022395 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022383 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022369 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022384 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022373 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022371 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022392 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022400 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022388 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022393 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022375 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022402 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022382 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022408 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022368 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022387 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022381 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26022378 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P146075 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P158677 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06579456 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01784284 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P273538 |
AK005954.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
AK082833.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
AK167332.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
NR_031759.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
BC094628.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
AK005954.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
AK082833.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
AK167332.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
NR_031759.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P273538 |
BC094628.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104873 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P146075 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P158677 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07400832 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06723264 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07187935 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07400833 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07187934 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26022400 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26022402 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26022398 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022490 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022459 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022466 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022497 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022409 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022437 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022461 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022346 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022395 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022457 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022463 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022499 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022420 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022473 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022432 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022501 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022413 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022505 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022388 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022441 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022400 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022507 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022471 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022422 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022517 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022387 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022405 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022390 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022383 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022502 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022485 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022483 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022454 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022509 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022511 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022447 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022460 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022416 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022392 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022342 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022478 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022408 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022510 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022513 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022385 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022424 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022465 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022455 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022431 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022464 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022515 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022402 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022397 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022417 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022448 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022418 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022399 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022492 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022481 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26022386 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022405 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022422 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022432 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022385 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022415 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022384 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022401 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022424 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06801272 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022436 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022392 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07400833 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022386 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022408 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022420 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022440 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06723264 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022389 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022442 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022388 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022406 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07426017 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022428 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022395 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07345633 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022402 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022404 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07187935 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022431 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07426021 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022383 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022409 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022426 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022429 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022399 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022387 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022393 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022441 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022394 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06824258 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07036840 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022417 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26022413 |
- |
- |