| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10443354 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443353 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443350 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443347 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443352 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443348 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443355 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443356 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443358 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443349 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443351 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10443359 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1453689_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1424197_PM_s_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10443346 |
- |
- |
| Mouse Genome 430 2.0 Array |
1424197_PM_s_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453689_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1424197_s_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1453689_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
72775_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
72775_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004127.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004137.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004103.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004134.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004111.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004111.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004111.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004111.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004111.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004111.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004111.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004113.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004115.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004102.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004104.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004106.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004107.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004117.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004117.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004117.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004117.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004117.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004130.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004130.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004130.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004130.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004130.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004118.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004123.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004135.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004136.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004105.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004114.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004119.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
NM_001163819.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
NM_001163819.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
NM_001163820.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
NM_001163820.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
NR_028296.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
NR_028296.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
NR_028297.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
NR_028297.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700004133.mm.1 |
TC1700000476.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1424197_s_at |
- |
- |
| Mouse Expression Set 430 |
1424197_PM_s_at |
- |
- |
| Mouse Expression Set 430 |
1453689_at |
- |
- |
| Mouse Expression Set 430 |
1453689_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1424197_s_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1424197_PM_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1424197_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1424197_PM_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453689_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1453689_PM_at |
- |
- |