Search
Search

Brca1| Gene Symbol: Brca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009764.3 | NP_033894.3 | 19 - 20 | 5311 | 90 | |
| GenBank mRNA | AB221610.1 | - | 10 - 11 | 2540 | 90 | |
| BC068303.1 | - | 19 - 20 | 5144 | 30431022 | 90 | |
| U31625.1 | - | 19 - 20 | 5168 | 90 | ||
| U32446.1 | - | 19 - 20 | 5221 | 90 | ||
| U35641.1 | - | 19 - 20 | 5204 | 90 | ||
| U36475.1 | - | 18 - 19 | 5131 | 90 | ||
| U68174.1 | - | 19 - 20 | 5215 | 90 | ||
Set Membership: |
|