| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100008012.mm.1 |
NM_016887.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008012.mm.1 |
NM_016887.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008012.mm.1 |
TC1100000932.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008014.mm.1 |
NM_016887.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008014.mm.1 |
NM_016887.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008014.mm.1 |
TC1100000932.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008017.mm.1 |
NM_001193619.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008017.mm.1 |
NM_001193619.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008017.mm.1 |
NM_016887.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008017.mm.1 |
NM_016887.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008017.mm.1 |
TC1100000932.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008013.mm.1 |
NM_016887.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008013.mm.1 |
NM_016887.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008013.mm.1 |
TC1100000932.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100008018.mm.1 |
NM_001193619.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008018.mm.1 |
NM_001193619.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008018.mm.1 |
NM_016887.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008018.mm.1 |
NM_016887.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100008018.mm.1 |
TC1100000932.mm.1 |
MTA TC |