| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30350209 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30350223 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30350224 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30350208 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
BC026997.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
AK163615.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
AK165937.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
BC019462.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
AK085641.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
BC026997.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
AK163615.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
AK165937.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
BC019462.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P215539 |
AK085641.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
645381 |
AK020485.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
645381 |
XM_125627.6 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
645381 |
XM_911116.3 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
112857 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
BC026997.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
AK163615.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
AK165937.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
BC019462.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
AK085641.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
BC026997.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
AK163615.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
AK165937.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
BC019462.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P215539 |
AK085641.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350238 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350274 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350257 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350253 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350271 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350226 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350258 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350270 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350259 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350237 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350224 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350265 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350205 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350239 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350204 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350229 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350256 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350261 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350246 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350206 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350251 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350227 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350250 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350236 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350209 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350240 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350245 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350230 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350273 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350233 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350242 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350247 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350267 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350223 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30350208 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P423143 |
BC026997.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P423143 |
AK163615.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P423143 |
AK165937.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P423143 |
AK193021.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P423143 |
BC019462.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
BC026997.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
AK163615.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
AK165937.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
BC019462.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
AK085641.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
BC026997.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
AK163615.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
AK165937.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
BC019462.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P215539 |
AK085641.1 |
mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12180192 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30350224 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30350208 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30350209 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30350223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350208 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350170 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350190 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350159 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350164 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350210 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350169 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350162 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350167 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350182 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350150 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350152 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350145 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350157 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350186 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350174 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350173 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350187 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350165 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350224 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350149 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350137 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350181 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350140 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350194 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350189 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30350168 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30350224 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30350223 |
- |
- |