Search
Search

Atp1a1| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144900.2 | NP_659149.1 | 17 - 18 | 2743 | 62 | |
| GenBank mRNA | AK088000.1 | - | 17 - 18 | 2704 | 62 | |
| AK137001.1 | - | 17 - 18 | 2703 | 62 | ||
| AK148989.1 | - | 4 - 5 | 601 | 62 | ||
| AK150523.1 | - | 10 - 11 | 1648 | 62 | ||
| AK159279.1 | - | 17 - 18 | 2700 | 62 | ||
| AK159306.1 | - | 17 - 18 | 2698 | 62 | ||
| AK191797.1 | - | 1 - 2 | 97 | 62 | ||
| AK192395.1 | - | 1 - 2 | 97 | 62 | ||
| AK192706.1 | - | 196 | 62 | |||
| AK193322.1 | - | 1 - 2 | 97 | 62 | ||
| AK195171.1 | - | 1 - 2 | 97 | 62 | ||
| AK196676.1 | - | 1 - 2 | 97 | 62 | ||
| AK197178.1 | - | 1 - 2 | 97 | 62 | ||
| AK197377.1 | - | 1 - 2 | 97 | 62 | ||
| AK197675.1 | - | 1 - 2 | 97 | 62 | ||
| AK199023.1 | - | 1 - 2 | 97 | 62 | ||
| AK199227.1 | - | 1 - 2 | 97 | 62 | ||
| AK200318.1 | - | 1 - 2 | 97 | 62 | ||
| AK201233.1 | - | 1 - 2 | 97 | 62 | ||
| AK202370.1 | - | 1 - 2 | 98 | 62 | ||
| AK202652.1 | - | 1 - 2 | 97 | 62 | ||
| AK203618.1 | - | 1 - 2 | 96 | 62 | ||
| AK204336.1 | - | 1 - 2 | 97 | 62 | ||
| AK204611.1 | - | 1 - 2 | 97 | 62 | ||
| AK204844.1 | - | 1 - 2 | 97 | 62 | ||
| AK205182.1 | - | 1 - 2 | 97 | 62 | ||
| BC010319.1 | - | 15 - 16 | 2328 | 62 | ||
| BC021496.1 | - | 17 - 18 | 2693 | 5346159 | 62 | |
| BC023175.1 | - | 1 - 2 | 37 | 62 | ||
| BC023794.1 | - | 18 - 19 | 2638 | 5351943 | 62 | |
| BC023964.1 | - | 17 - 18 | 2684 | 62 | ||
| BC025037.1 | - | 10 - 11 | 1543 | 62 | ||
| BC025618.1 | - | 17 - 18 | 2702 | 5320976 | 62 | |
| BC025627.1 | - | 17 - 18 | 2693 | 5320513 | 62 | |
| BC025811.1 | - | 17 - 18 | 2680 | 5096872 | 62 | |
| BC030326.1 | - | 6 - 7 | 1040 | 62 | ||
| BC031176.1 | - | 4 - 5 | 601 | 62 | ||
| BC031389.1 | - | 1 - 2 | 162 | 62 | ||
| BC032187.1 | - | 17 - 18 | 2743 | 5321268 | 62 | |
| BC033435.1 | - | 17 - 18 | 2639 | 4950746 | 62 | |
| BC033471.1 | - | 17 - 18 | 2683 | 4954119 | 62 | |
| BC034177.1 | - | 16 - 17 | 2448 | 62 | ||
| BC035352.1 | - | 16 - 17 | 2413 | 62 | ||
| BC042435.1 | - | 17 - 18 | 2693 | 4951288 | 62 | |
Set Membership: |
|