Search
Search

Olfr642| Gene Symbol: Olfr642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146329.1 | NP_666441.1 | 1 | 48 | 65 | |
| GenBank mRNA | BC126862.1 | - | 1 | 48 | 40042372 | 65 |
| BC126863.1 | - | 1 | 48 | 40042373 | 65 | |
Set Membership: |
|