Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00527554_s1
          See other OLFR648 GE Assays ›
          Gene Symbol:
          Olfr648
          Gene Name:
          olfactory receptor 648
          Gene Aliases:
          MOR31-12
          Chromosome Location:
          Chr.7: 104179456 - 104180406 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00527554_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr648
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146751.1 NP_666962.1 1 258 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003932.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr648

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2104174 NM_146751.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2104174 BC106984.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112603 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2104174 NM_146751.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2104174 BC106984.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398859 NM_146751.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398859 BC106984.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25386999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25387041 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112603 - -
          Whole Mouse Genome G4122A A_51_P398859 NM_146751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398859 BC106984.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103552 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112603 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06077018 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06459213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06077019 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06459223 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06459219 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06459229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25386986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25387003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06459215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06459228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06357735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25387002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06172470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25386994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06229148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06288335 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566323 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566322 - -
          Affymetrix Mouse Clariom S Array PSR0700032863.mm.1 NM_146751.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032863.mm.1 NM_146751.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032863.mm.1 TC0700003932.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 440353 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3870097 - -
          Illumina mouse-6 v1.0 expression beadchip 3870097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2688825 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2688825 - -
          mouse-6 v1.1 (Illumina) scl8861.1.1_279-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8861.1.1_279-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256392 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256392 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0