| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
NM_001130184.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
NM_001130184.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
NM_001130185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
NM_001130185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
NM_001130186.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
NM_001130186.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
NM_172303.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
NM_172303.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002395.mm.1 |
TC0300000322.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
NM_001130184.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
NM_001130184.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
NM_001130185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
NM_001130185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
NM_001130186.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
NM_001130186.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
NM_172303.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
NM_172303.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002396.mm.1 |
TC0300000322.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300002392.mm.1 |
NM_001130185.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002392.mm.1 |
NM_001130185.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002392.mm.1 |
TC0300000322.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300002393.mm.1 |
NM_001130186.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002393.mm.1 |
NM_001130186.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300002393.mm.1 |
TC0300000322.mm.1 |
MTA TC |