| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200006792.mm.1 |
NM_175238.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006792.mm.1 |
TC0200000821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200006794.mm.1 |
NM_175238.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006794.mm.1 |
TC0200000821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200006793.mm.1 |
NM_175238.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006793.mm.1 |
TC0200000821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200006801.mm.1 |
NM_175238.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006801.mm.1 |
TC0200000821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200006796.mm.1 |
NM_175238.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006796.mm.1 |
TC0200000821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200006797.mm.1 |
NM_175238.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006797.mm.1 |
TC0200000821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200006798.mm.1 |
NM_175238.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006798.mm.1 |
TC0200000821.mm.1 |
MTA TC |