Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00615218_gH
          See other ZFP710 GE Assays ›
          Gene Symbol:
          Zfp710
          Gene Name:
          zinc finger protein 710
          Gene Aliases:
          5430400N05Rik, 9830165K13, AI324798, Znf710
          Chromosome Location:
          Chr.7: 80024528 - 80092751 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00615218_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp710
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145999.1 NP_001139471.1 3 - 4 1862 64
          NM_001146000.1 NP_001139472.1 3 - 4 1862 64
          NM_175433.5 NP_780642.3 3 - 4 1945 64
          GenBank mRNA AK036711.1 - 2 - 3 520 64
          AK083979.1 - 3 - 4 1787 64
          AK089457.1 - 2 - 3 1137 64
          AK154871.1 - 3 - 4 1843 64
          AK154953.1 - 3 - 4 1863 64
          AK155416.1 - 3 - 4 1924 64
          BC058640.1 - 3 - 4 1756 6835601 64
          BC060641.1 - 3 - 4 1839 6849964 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001141.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp710

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          biological_process
          nucleic acid binding
          DNA binding
          transcription cofactor activity
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700010009.mm.1 TC0700001141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010009.mm.1 NM_001145999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010009.mm.1 NM_001145999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010009.mm.1 NM_001146000.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010009.mm.1 NM_001146000.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010012.mm.1 TC0700001141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010012.mm.1 NM_175433.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010012.mm.1 NM_175433.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010011.mm.1 TC0700001141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010011.mm.1 NM_175433.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010011.mm.1 NM_175433.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010017.mm.1 TC0700001141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010017.mm.1 NM_001146000.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010017.mm.1 NM_001146000.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010013.mm.1 TC0700001141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010013.mm.1 NM_001145999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010013.mm.1 NM_001145999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010013.mm.1 NM_001146000.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010013.mm.1 NM_001146000.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010013.mm.1 NM_175433.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010013.mm.1 NM_175433.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010015.mm.1 TC0700001141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010015.mm.1 NM_001145999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010015.mm.1 NM_001145999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010015.mm.1 NM_001146000.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010015.mm.1 NM_001146000.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010015.mm.1 NM_175433.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010015.mm.1 NM_175433.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010010.mm.1 TC0700001141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010010.mm.1 NM_175433.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010010.mm.1 NM_175433.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010014.mm.1 TC0700001141.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010014.mm.1 NM_001145999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010014.mm.1 NM_001145999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010014.mm.1 NM_001146000.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010014.mm.1 NM_001146000.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010014.mm.1 NM_175433.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010014.mm.1 NM_175433.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 362079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline