Search
Search

Ccdc88a| Gene Symbol: Ccdc88a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176841.4 | NP_789811.2 | 2 - 3 | 339 | 131 | |
| GenBank mRNA | AB087827.1 | - | 2 - 3 | 163 | 131 | |
| AK051140.1 | - | 2 - 3 | 737 | 131 | ||
Set Membership: |
|