Search
Search

Ccdc69| Gene Symbol: Ccdc69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177471.4 | NP_803422.2 | 3 - 4 | 279 | 58 | |
| GenBank mRNA | AK008760.1 | - | 3 - 4 | 250 | 58 | |
| AK079810.1 | - | 3 - 4 | 280 | 58 | ||
| AK135942.1 | - | 3 - 4 | 256 | 58 | ||
| AK170686.1 | - | 3 - 4 | 229 | 58 | ||
| BC096763.1 | - | 3 - 4 | 237 | 30053993 | 58 | |
Set Membership: |
|