Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00625888_m1
          See other C130026L21RIK GE Assays ›
          Gene Symbol:
          C130026L21Rik
          Gene Name:
          RIKEN cDNA C130026L21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 111581561 - 111587153 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00625888_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C130026L21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015546.2 3 - 4 513 108
          GenBank mRNA AK047985.1 - 355 108
          AK160717.1 - 3 - 4 330 108
          AK160721.1 - 3 - 4 514 108
          BC094571.1 - 2 - 3 190 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA NM_177875.1 - 355 Chr. - - [-] on Build 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001168.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C130026L21Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030020 BC094571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030020 AK047985.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030020 AK160721.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030020 NR_015546.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416568 AK016003.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155900 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176395 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05696360 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176395 - -
          Whole Mouse Genome G4122A A_51_P330537 AK047985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P276869 AK038902.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04549375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04656985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04621297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04246392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04226019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04268210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04606682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04593589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04814856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04593586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04830353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04656772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04912789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04694619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04593949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04593587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04247810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04452859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04549386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04621296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04656785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04268205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04268214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04912790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04783714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04712582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04549379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04912794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04656979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04831472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04273714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04458183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04147211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04147214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04549383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04593595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04830351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04912797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04247812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04549377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04965408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04656976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04549376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04273713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04276366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04593588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04512241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04610767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04226024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04912795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04226021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04656977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04246393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04769039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04881318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04549374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04226017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23986828 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10524336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10524335 - -
          Affymetrix HT MG-430 PM Array Plate 1442868_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10524331 - -
          Mouse Genome 430 2.0 Array 1442868_PM_at - -
          Mouse Genome 430 2.0 Array 1442868_at - -
          Affymetrix Mouse Clariom S Array PSR0500009996.mm.1 TC0500001168.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500009996.mm.1 NR_015546.2 Refseq
          Mouse Expression Set 430 1442868_at - -
          Mouse Expression Set 430 1442868_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442868_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442868_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 343917 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100510301 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline