| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345383 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345342 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345341 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345385 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345373 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345339 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345381 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345378 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345376 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345375 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345340 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345382 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345377 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345384 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345380 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P136778 |
AK153712.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P136778 |
NM_177922.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P136778 |
BC048082.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
513141 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK004703.2 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK028817.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK033823.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK037059.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK037127.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK038944.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK042469.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK128959.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
AK167155.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
BC026837.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
BC066833.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
696169 |
NM_027890.4 |
refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P120166 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P136778 |
AK153712.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P136778 |
NM_177922.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P136778 |
BC048082.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345351 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345346 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345335 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345344 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345361 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345357 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345334 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345340 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345360 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345356 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345355 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345353 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345348 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345345 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345328 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345330 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345337 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345327 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345338 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345333 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345350 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30345358 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08420867 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P120166 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P540365 |
NM_177922.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P540365 |
BC048082.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P540360 |
BC053920.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P540360 |
NM_177922.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P540360 |
BC048082.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136778 |
AK153712.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136778 |
NM_177922.2 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136778 |
BC048082.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125310 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P120166 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11904297 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11904305 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12356946 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11904299 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11904303 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12356945 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345376 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345378 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345375 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345381 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345342 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345383 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345340 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345384 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345380 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345373 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345385 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345339 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345341 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345382 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30345377 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345385 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345299 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345313 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345388 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345447 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345344 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345325 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345360 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345373 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345306 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345334 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345446 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345358 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345455 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345371 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345350 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345322 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345384 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345381 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345460 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345287 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345316 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345437 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345345 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345389 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345386 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345378 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345377 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345380 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345296 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345445 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345308 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345333 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345461 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345364 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345470 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345335 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345443 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345468 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345475 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345456 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345348 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345303 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345301 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345365 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345394 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345331 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345476 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345310 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345355 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345463 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345444 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345458 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345449 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345453 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345477 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345441 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345340 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345391 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345351 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345473 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345346 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345375 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345338 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345452 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345288 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345465 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345353 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345362 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345361 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345369 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345439 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30345356 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345308 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12387939 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12139579 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345286 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345310 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345287 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345350 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12302243 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12176515 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345345 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345320 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345386 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345297 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345351 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345391 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12378065 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12356946 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345383 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345369 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345335 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345344 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345319 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345355 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12176525 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345348 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345364 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345362 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12176507 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345288 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345380 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345365 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345293 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345303 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12356945 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345366 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345357 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345371 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345358 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345361 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11871094 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345333 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345330 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345377 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345325 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11935056 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12437037 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345309 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345394 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345373 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12045508 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345340 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345299 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345381 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345291 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345304 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345346 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11904305 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345328 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345356 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345301 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345360 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30345292 |
- |
- |