Search
Search

Nrcam| Gene Symbol: Nrcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146031.1 | NP_001139503.1 | 20 - 21 | 2800 | 62 | |
| NM_176930.4 | NP_795904.3 | 21 - 22 | 2830 | 62 | ||
| GenBank mRNA | AJ543321.1 | - | 21 - 22 | 2732 | 62 | |
| AK087693.1 | - | 20 - 21 | 2714 | 62 | ||
| AK122252.1 | - | 20 - 21 | 2730 | 62 | ||
| BC055053.1 | - | 20 - 21 | 2445 | 62 | ||
Set Membership: |
|