| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969962 |
AY263400.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969962 |
BC089518.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1969962 |
HM179540.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P125729 |
NM_029772.3 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P125729 |
AK014942.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
857282 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729991 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
675625 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429659 |
AK018094.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429659 |
AK042761.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429659 |
AK044973.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429659 |
AK047610.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429659 |
NM_029529.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
407524 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
114083 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757920 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
795817 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
374489 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
502301 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
105 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
836542 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
589885 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
659028 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
262586 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
150536 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
54519 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84972 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
86152 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P115149 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169313 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969962 |
AY263400.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969962 |
BC089518.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1969962 |
HM179540.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P125729 |
NM_029772.3 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P125729 |
AK014942.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P246233 |
HM179541.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P246233 |
NM_029772.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P246233 |
AK014942.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P246233 |
AY263400.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P246233 |
BC089518.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057029 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057024 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057025 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057026 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057030 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057015 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057020 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057022 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057041 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057048 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057043 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057035 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057039 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057033 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057018 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057016 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057038 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057028 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057045 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057036 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057031 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29057034 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02697609 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02697625 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169313 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07883841 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P115149 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P246233 |
HM179541.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P246233 |
NM_029772.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P246233 |
AK014942.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P246233 |
AY263400.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P246233 |
BC089518.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139600 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125729 |
NM_029772.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125729 |
AK014942.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169313 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P02697625 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P115149 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057114 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057085 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057043 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057000 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057035 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057206 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057125 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057025 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057186 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057177 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057189 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057031 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057179 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057039 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057100 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057087 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057167 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057104 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057133 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057136 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057076 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057158 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057110 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057030 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057007 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057141 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057016 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057116 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057012 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057059 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057009 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057159 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057092 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057003 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057121 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057089 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057165 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057015 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057142 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057182 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057036 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057068 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057148 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057111 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057173 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057073 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057138 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057095 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057033 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057152 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057150 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057131 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057109 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057127 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057078 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057187 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057194 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057204 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057129 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057070 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29056998 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057079 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057108 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057045 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057060 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29056994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057140 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057034 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057041 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057106 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057112 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29057145 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10934551 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057026 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057161 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057012 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057035 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10781461 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057171 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057179 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057007 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057048 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057039 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057002 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057185 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057140 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057184 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057000 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057005 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057182 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10843346 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057030 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057145 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057014 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057033 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057057 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057009 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10781466 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057167 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057158 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057062 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057059 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057015 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057031 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10963799 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057053 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057020 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10817962 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057024 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057010 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057061 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057186 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10660404 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057036 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057189 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10958164 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057159 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057147 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10580572 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057192 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11025988 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11107938 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10716916 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10958165 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057025 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10589019 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10892674 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057177 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29056996 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057041 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057016 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057018 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10892675 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10958166 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29056998 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057045 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057165 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057043 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057150 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P29057148 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10892681 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10781455 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10727246 |
- |
- |