Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00729950_s1
          See other OLFR315 GE Assays ›
          Gene Symbol:
          Olfr315
          Gene Name:
          olfactory receptor 315
          Gene Aliases:
          MOR278-1
          Chromosome Location:
          Chr.11: 58778089 - 58779086 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00729950_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr315
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146538.2 NP_666749.1 1 390 139
          GenBank mRNA BC119148.1 - 1 390 8733897 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr315

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P643087 NM_146538.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P643087 BC119148.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P643087 NM_146538.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P643087 BC119148.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P643087 NM_146538.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P643087 BC119148.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P643087 NM_146538.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P643087 BC119148.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P643087 NM_146538.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P643087 BC119148.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P643087 NM_146538.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P643087 BC119148.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27832693 - -
          Whole Mouse Genome G4122A A_52_P643087 NM_146538.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643087 BC119148.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643087 NM_146538.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643087 BC119148.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09717132 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09483850 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09033026 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09717142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09717137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09038961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09463538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09693219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09182117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09033027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09487164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09618528 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376411 - -
          Affymetrix HT MG-430 PM Array Plate 1445219_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10376410 - -
          Mouse Genome 430 2.0 Array 1445219_PM_at - -
          Mouse Genome 430 2.0 Array 1445219_at - -
          Mouse Expression Set 430 1445219_at - -
          Mouse Expression Set 430 1445219_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445219_at - -
          Affymetrix HT Mouse Genome 430B Array 1445219_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 811905 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3870053 - -
          Illumina mouse-6 v1.0 expression beadchip 3870053 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870053 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2700931 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2700931 - -
          mouse-6 v1.1 (Illumina) scl41515.1.1_98-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870053 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41515.1.1_98-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2700931 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2700931 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0