| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900025242.mm.1 |
NM_153413.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025242.mm.1 |
NM_153413.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025242.mm.1 |
TC0900003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025243.mm.1 |
NM_153413.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025243.mm.1 |
NM_153413.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025243.mm.1 |
TC0900003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025245.mm.1 |
NM_153413.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025245.mm.1 |
NM_153413.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025245.mm.1 |
TC0900003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025246.mm.1 |
NM_153413.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025246.mm.1 |
NM_153413.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025246.mm.1 |
TC0900003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025248.mm.1 |
NM_153413.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025248.mm.1 |
NM_153413.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025248.mm.1 |
TC0900003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025241.mm.1 |
NM_153413.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025241.mm.1 |
NM_153413.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025241.mm.1 |
TC0900003023.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900025244.mm.1 |
NM_153413.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025244.mm.1 |
NM_153413.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900025244.mm.1 |
TC0900003023.mm.1 |
MTA TC |