Search
Search
| Gene Symbol: Cars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252593.1 | NP_001239522.1 | 17 - 18 | 1909 | 63 | |
| NM_013742.5 | NP_038770.3 | 18 - 19 | 2158 | 63 | ||
| GenBank mRNA | AJ276796.1 | - | 18 - 19 | 2125 | 63 | |
| AK033328.1 | - | 18 - 19 | 2125 | 63 | ||
| AK037227.1 | - | 17 - 18 | 1877 | 63 | ||
| AK047027.1 | - | 17 - 18 | 1877 | 63 | ||
| AK077558.1 | - | 17 - 18 | 1875 | 63 | ||
| AK133205.1 | - | 17 - 18 | 1876 | 63 | ||
| AK152866.1 | - | 17 - 18 | 1882 | 63 | ||
| BC030473.1 | - | 14 - 15 | 1594 | 63 | ||
| BC054717.1 | - | 18 - 19 | 2107 | 5686364 | 63 | |
| BC056194.1 | - | 4 - 5 | 460 | 63 | ||
| BC058954.1 | - | 18 - 19 | 2110 | 5707198 | 63 | |
Set Membership: |
|