Search
Search

Srebf1| Gene Symbol: Srebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011480.3 | NP_035610.1 | 5 - 6 | 1110 | 80 | |
| GenBank mRNA | AB017337.1 | - | 4 - 5 | 926 | 80 | |
| AF374266.1 | - | 4 - 5 | 874 | 80 | ||
| AK052628.1 | - | 5 - 6 | 1111 | 80 | ||
| AK150052.1 | - | 5 - 6 | 1099 | 80 | ||
| AK154424.1 | - | 5 - 6 | 1078 | 80 | ||
| AK169607.1 | - | 5 - 6 | 1111 | 80 | ||
| BC006051.1 | - | 2 - 3 | 246 | 80 | ||
| BC056922.1 | - | 5 - 6 | 1092 | 6824948 | 80 | |
Set Membership: |
|