Search
Search

Snhg4| Gene Symbol: Snhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038073.1 | 2 - 3 | 94 | 92 | ||
| GenBank mRNA | AK041908.1 | - | 2 - 3 | 95 | 92 | |
| AK080350.1 | - | 2 - 3 | 62 | 92 | ||
| AK088959.1 | - | 2 - 3 | 92 | 92 | ||
Set Membership: |
|