Search
Search

Map6d1| Gene Symbol: Map6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198599.2 | NP_941001.2 | 1 - 2 | 421 | 69 | |
| GenBank mRNA | AK079603.1 | - | 1 - 2 | 384 | 69 | |
| AK163656.1 | - | 1 - 2 | 385 | 69 | ||
| BB653005.1 | - | 1 - 2 | 422 | 69 | ||
| BC116220.1 | - | 1 - 2 | 384 | 40091320 | 69 | |
| BC116221.1 | - | 1 - 2 | 384 | 40091321 | 69 | |
| BE979311.1 | - | 1 - 2 | 95 | 69 | ||
Set Membership: |
|