Search
Search

Dscaml1| Gene Symbol: Dscaml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081270.1 | NP_001074739.1 | 1 - 2 | 221 | 57 | |
| GenBank mRNA | AF487345.1 | - | 1 - 2 | 125 | 57 | |
| AF487347.1 | - | 1 - 2 | 113 | 57 | ||
| AK147531.1 | - | 1 - 2 | 165 | 57 | ||
Set Membership: |
|